missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylene Blue, 1% in Ethanol, Certified, LabChem™
Specifications
Specifications
| Chemical Name or Material | Methylene Blue |
| Absorbance | Passes Test |
| Melting Point | -115°C |
| Name Note | 1% in Ethanol |
| Boiling Point | 78°C |
| CAS Max % | 89.1, 4.95, 4.95, 1 |
| CAS | 67-63-0, 67-56-1, 61-73-4 |
| Linear Formula | C16H18N3SCl |
| Solubility Information | Soluble in water |
| Identification | Passes Test |
| Show More |