missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylene Blue, 0.1% (w/v), Ricca Chemical
Supplier: Ricca Chemical Company 484132
Specifications
| Methylene Blue | |
| 0.1% W/V | |
| 0.1% (w/v) Aqueous Solution | |
| Blue |
| 7732-18-5,7220-79-3 | |
| Laboratory | |
| Amber Glass Bottle | |
| 1 L |