missing translation for 'onlineSavingsMsg'
Learn More

Methyle5-methyl-3-isoxazolecarboxylate, 97%, Thermo Scientific™

Catalog No. CC10422DA
Change view
Click to view available options
Quantity:
1g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC10422DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC10422DA Supplier Thermo Scientific Chemicals Supplier No. CC10422DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19788-35-3
Molecular Formula C6H7NO3
Molecular Weight (g/mol) 141.13
MDL Number MFCD00015895
InChI Key MVHHQOCEOUNTID-UHFFFAOYSA-N
Synonym methyl 5-methylisoxazole-3-carboxylate, methyl 5-methyl-3-isoxazolecarboxylate, 3-isoxazolecarboxylic acid, 5-methyl-, methyl ester, 5-methyl-3-isoxazolecarboxylic acid methyl ester, methyl 5-methyl-3-isoxazole carboxylate, methyl-5-methyl-3-isoxazole carboxylate, 5-methylisoxazole-3-carboxylic acid methyl ester, 5-methyl-isoxazole-3-carboxylic acid methyl ester, acmc-209xf5, ksc489s1r
PubChem CID 88245
IUPAC Name methyl 5-methyl-1,2-oxazole-3-carboxylate
SMILES COC(=O)C1=NOC(C)=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 141.13
Percent Purity 97%
Chemical Name or Material Methyle5-methyl-3-isoxazolecarboxylate