missing translation for 'onlineSavingsMsg'
Learn More

Methyle5-(chlorosulfonyl)-4-methyl-2-thiophenecarboxylate, 97%, Thermo Scientific™

Chemical Identifiers

CAS 423768-46-1
Molecular Formula C7H7ClO4S2
Molecular Weight (g/mol) 254.699
MDL Number MFCD03407367
InChI Key YWBXGHFHMHUSKH-UHFFFAOYSA-N
Synonym methyl 5-chlorosulfonyl-4-methylthiophene-2-carboxylate, methyl 5-chlorosulfonyl-4-methyl-2-thiophenecarboxylate, 2-thiophenecarboxylicacid, 5-chlorosulfonyl-4-methyl-, methyl ester
PubChem CID 2779815
IUPAC Name methyl 5-chlorosulfonyl-4-methylthiophene-2-carboxylate
SMILES CC1=C(SC(=C1)C(=O)OC)S(=O)(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
MO00640CB
View Documents
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

423768-46-1
254.699
YWBXGHFHMHUSKH-UHFFFAOYSA-N
2779815
CC1=C(SC(=C1)C(=O)OC)S(=O)(=O)Cl
C7H7ClO4S2
MFCD03407367
methyl 5-chlorosulfonyl-4-methylthiophene-2-carboxylate, methyl 5-chlorosulfonyl-4-methyl-2-thiophenecarboxylate, 2-thiophenecarboxylicacid, 5-chlorosulfonyl-4-methyl-, methyl ester
methyl 5-chlorosulfonyl-4-methylthiophene-2-carboxylate

Specifications

423768-46-1
97%
MFCD03407367
YWBXGHFHMHUSKH-UHFFFAOYSA-N
methyl 5-chlorosulfonyl-4-methylthiophene-2-carboxylate
2779815
97%
97.0
C7H7ClO4S2
methyl 5-chlorosulfonyl-4-methylthiophene-2-carboxylate, methyl 5-chlorosulfonyl-4-methyl-2-thiophenecarboxylate, 2-thiophenecarboxylicacid, 5-chlorosulfonyl-4-methyl-, methyl ester
CC1=C(SC(=C1)C(=O)OC)S(=O)(=O)Cl
254.699
254.71
Methyl 5-(chlorosulfonyl)-4-methyl-2-thiophenecarboxylate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents