missing translation for 'onlineSavingsMsg'
Learn More

Methylcyclopentane 96.0+%, TCI America™

Catalog No. M020325ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M020325ML 25 mL
M0203500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M020325ML Supplier TCI America Supplier No. M020325ML

Chemical Identifiers

CAS 96-37-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.162
MDL Number MFCD00001382
InChI Key GDOPTJXRTPNYNR-UHFFFAOYSA-N
Synonym cyclopentane, methyl, methyl cyclopentane, methylpentamethylene, unii-5g26cc1ask, methyl-cyclopentane, ccris 6058, hsdb 876, 5g26cc1ask, un2298 flammable liquid, 3-methylcyclopentyl
PubChem CID 7296
IUPAC Name methylcyclopentane
SMILES CC1CCCC1

Specifications

Color Colorless
Boiling Point 72°C
Quantity 25 mL
UN Number 2298
Formula Weight 84.16
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Methylcyclopentane
RTECSNumber GY4640000
TSCA Yes