missing translation for 'onlineSavingsMsg'
Learn More

Methylcyclopentane, 95%

Catalog No. p-162277
Change view
Click to view available options
Quantity:
500 mL
2.5 L
25 L
Packaging:
Glass bottle
Metal Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC345245000 500 mL Glass bottle
AC345240025 2.5 L Glass bottle
AC345240250 25 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC345245000 Supplier Thermo Scientific Chemicals Supplier No. 345245000
Only null left

CAS: 96-37-7 | C6H12 | 84.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 96-37-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00001382
InChI Key GDOPTJXRTPNYNR-UHFFFAOYSA-N
Synonym cyclopentane, methyl, methyl cyclopentane, methylpentamethylene, unii-5g26cc1ask, methyl-cyclopentane, ccris 6058, hsdb 876, 5g26cc1ask, un2298 flammable liquid, 3-methylcyclopentyl
PubChem CID 7296
IUPAC Name methylcyclopentane
SMILES CC1CCCC1

Specifications

Melting Point -142.4°C
Color White
Density 0.7480g/mL
Boiling Point 71.0°C
Flash Point −4°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4077 to 1.4097
Linear Formula C5H9CH3
Quantity 500 mL
Beilstein 05, 27
Specific Gravity 0.748
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone,benzene,carbon tetrachloride,,diethyl ether,ethanol,petroleum ether
Viscosity 0.505 mPa.s (20°C)
Formula Weight 84.16
Percent Purity 95%
Physical Form Adhering Crystals
Chemical Name or Material Methylcyclopentane, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
May be fatal if swallowed and enters airways.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-503-2
RTECSNumber GY4640000
TSCA TSCA

RUO – Research Use Only