missing translation for 'onlineSavingsMsg'
Learn More

Methylcyclopentadiene dimer, tech. 90%, stab. with 200ppm 4-tert-butylcatechol

Catalog No. AAA17596AU
Change view
Click to view available options
Quantity:
500 mL
1000 mL
5000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA17596AU 1000 mL
AAA17596AP 500 mL
AAA175960J 5000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA17596AU Supplier Thermo Scientific Chemicals Supplier No. A17596AU

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 26472-00-4 | C12H16

Methylcyclopentadiene dimer is used to study the competition with ethylene for binding and biological activities. Used in the synthesis of rubbers and resins intended for use in contact with food or drink.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylcyclopentadiene dimer is used to study the competition with ethylene for binding and biological activities. Used in the synthesis of rubbers and resins intended for use in contact with food or drink.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 26472-00-4
Molecular Formula C12H16
MDL Number MFCD00082297

Specifications

Melting Point ∼-50°C
Density 0.94
Boiling Point 198°C to 201°C
Flash Point 26°C (79°F)
Refractive Index 1.498
Quantity 1000 mL
UN Number UN3295
Solubility Information Insoluble in water.
Formula Weight 160.26
Percent Purity 90%
Grade Technical
Chemical Name or Material Methylcyclopentadiene dimer, stabilized with 200ppm 4-tert-butylcatechol
Show More Show Less
Hazard Category H226-H351
Hazard Statement GHS H Statement
H340-H350-H226
May cause genetic defects.
May cause cancer.
Flammable liquid and vapour.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P280-P308+P313-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 247-724-5
RTECSNumber PC1075000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only