missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylcyclohexene-1,2-dicarboxylic Anhydride (mixture of isomers) 80.0+%, TCI America™
$57.79 - $201.32
Chemical Identifiers
| CAS | 11070-44-3 |
|---|---|
| Molecular Formula | C9H10O3 |
| MDL Number | MFCD00014585 |
| Synonym | Methyltetrahydrophthalic Anhydride |
Chemical Identifiers
| 11070-44-3 | |
| C9H10O3 | |
| MFCD00014585 | |
| Methyltetrahydrophthalic Anhydride |
Specifications
| 11070-44-3 | |
| 120°C | |
| MFCD00014585 | |
| 3082 | |
| 166.18 | |
| Liquid |
| Yellow | |
| C9H10O3 | |
| 25 g | |
| Methyltetrahydrophthalic Anhydride | |
| ≥80.0% (GC) | |
| Methylcyclohexene-1,2-dicarboxylic Anhydride (mixture of isomers) |
Safety and Handling
EINECSNumber : (3)-2451&(3)-2479
RTECSNumber : TI3325000
TSCA : Yes