missing translation for 'onlineSavingsMsg'
Learn More

Methylcyclohexene-1,2-dicarboxylic Anhydride (mixture of isomers) 80.0+%, TCI America™

Catalog No. M0683500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0683500G 500 g
M068325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0683500G Supplier TCI America Supplier No. M0683500G

Chemical Identifiers

CAS 11070-44-3
Molecular Formula C9H10O3
MDL Number MFCD00014585
Synonym Methyltetrahydrophthalic Anhydride

Specifications

Color Yellow
Boiling Point 120°C
Quantity 500 g
UN Number 3082
Formula Weight 166.18
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material Methylcyclohexene-1,2-dicarboxylic Anhydride (mixture of isomers)
EINECSNumber (3)-2451&(3)-2479
RTECSNumber TI3325000
TSCA Yes