missing translation for 'onlineSavingsMsg'
Learn More

Methylcyclohexane 99.0+%, TCI America™

Catalog No. M0190500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0190500ML 500 mL
M019025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0190500ML Supplier TCI America Supplier No. M0190500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-87-2
Molecular Formula C7H14
Molecular Weight (g/mol) 98.19
MDL Number MFCD00001497
InChI Key UAEPNZWRGJTJPN-UHFFFAOYSA-N
Synonym cyclohexane, methyl, hexahydrotoluene, cyclohexylmethane, toluene hexahydride, hexahydroxytoluene, sextone b, toluene, hexahydro, metylocykloheksan, methyl cyclohexane, methylcyclohexan
PubChem CID 7962
IUPAC Name methylcyclohexane
SMILES CC1CCCCC1

Specifications

Melting Point -127°C
Color Colorless
Boiling Point 100°C
Quantity 500 mL
UN Number 2296
Formula Weight 98.19
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methylcyclohexane
EINECSNumber (3)-2230
RTECSNumber GV6125000
TSCA Yes