missing translation for 'onlineSavingsMsg'
Learn More

Methylcyclohexane, 99%

Catalog No. p-7023029
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA16057AE 100 mL
AAA16057AP 500 mL
AAA160570F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA16057AE Supplier Thermo Scientific Chemicals Supplier No. A16057AE
Only null left

CAS: 108-87-2 | C7H14 | 98.19 g/mol

Methylcyclohexane is used as a solvent and reagent in organic synthesis and polymer chemistry. It is a component of jet fuel and also used as correction fluid in certain consumer products. It acts as a starting material for toluene synthesis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylcyclohexane is used as a solvent and reagent in organic synthesis and polymer chemistry. It is a component of jet fuel and also used as correction fluid in certain consumer products. It acts as a starting material for toluene synthesis

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 108-87-2
Molecular Formula C7H14
Molecular Weight (g/mol) 98.19
MDL Number MFCD00001497
InChI Key UAEPNZWRGJTJPN-UHFFFAOYSA-N
Synonym cyclohexane, methyl, hexahydrotoluene, cyclohexylmethane, toluene hexahydride, hexahydroxytoluene, sextone b, toluene, hexahydro, metylocykloheksan, methyl cyclohexane, methylcyclohexan
PubChem CID 7962
IUPAC Name methylcyclohexane
SMILES CC1CCCCC1

Specifications

Melting Point -126°C
Density 0.771
Boiling Point 101°C
Flash Point −4°C (−25°F)
Refractive Index 1.423
Quantity 100 mL
UN Number UN2296
Beilstein 505972
Merck Index 14,6047
Solubility Information Immiscible with water.
Formula Weight 98.19
Percent Purity 99%
Chemical Name or Material Methylcyclohexane
Show More Show Less
Hazard Category H225-H304-H315-H319-H335-H336
Hazard Statement GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHYLCYCLOHEXANE
EINECSNumber 203-624-3
RTECSNumber GV6125000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only