missing translation for 'onlineSavingsMsg'
Learn More

Methylcyclohexane, 98+%, Extra Dry, AcroSeal™

Catalog No. AC397241000
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC397241000 100 mL
AC397240010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC397241000 Supplier Thermo Scientific Chemicals Supplier No. 397241000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108-87-2 | C7H14 | 98.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108-87-2
Molecular Formula C7H14
Molecular Weight (g/mol) 98.19
MDL Number MFCD00001497
InChI Key UAEPNZWRGJTJPN-UHFFFAOYSA-N
Synonym cyclohexane, methyl, hexahydrotoluene, cyclohexylmethane, toluene hexahydride, hexahydroxytoluene, sextone b, toluene, hexahydro, metylocykloheksan, methyl cyclohexane, methylcyclohexan
PubChem CID 7962
IUPAC Name methylcyclohexane
SMILES CC1CCCCC1

Specifications

Melting Point -126.3°C
Color Colorless
Density 0.7700g/mL
Boiling Point 101.0°C
Flash Point −3°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging AcroSeal glass Bottle
Refractive Index 1.4210 to 1.4230
Linear Formula C6H11CH3
Quantity 100 mL
Residue after Evaporation 0.001% max.
Specific Gravity 0.77
Merck Index 15, 6118
Solubility Information Solubility in water: 0.1g/L (20°C). Other solubilities: soluble in acetone,benzene,carbon tetrachloride,,ether,ethanol
Formula Weight 98.19
Percent Purity 98+%
Grade Extra Dry
Physical Form Liquid
Chemical Name or Material Methylcyclohexane, Extra Dry
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-624-3

RUO – Research Use Only