missing translation for 'onlineSavingsMsg'
Learn More

Methylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. M15531G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M15531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M15531G Supplier TCI America Supplier No. M15531G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13061-96-6
Molecular Formula CH5BO2
MDL Number MFCD00002105
InChI Key KTMKRRPZPWUYKK-UHFFFAOYSA-N
PubChem CID 139377
IUPAC Name methylboronic acid

Specifications

Melting Point 91°C
Color White-Yellow
Quantity 1 g
Synonym methaneboronic acid, methyl boronic acid, dihydroxymethylborane, boronic acid, methyl, unii-8z1me41sch, methane boronic acid, 8z1me41sch, methylboronicacid, methyboronic acid, methyl-boronic acid
SMILES CB(O)O
Molecular Weight (g/mol) 59.86
Formula Weight 59.86
Physical Form Crystalline Powder
Chemical Name or Material Methylboronic Acid (contains varying amounts of Anhydride)