missing translation for 'onlineSavingsMsg'
Learn More

Methylammonium Tetrafluoroborate 98.0+%, TCI America™

Catalog No. M29901G
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
M29901G 1 g
M29905G 5 g
2 options

Catalog No. M29901G

Supplier: TCI America M29901G

Only null left

Chemical Identifiers

CAS 42539-74-2
Molecular Formula CH6BF4N
Molecular Weight (g/mol) 118.87
InChI Key ILEINUTYCFMMSF-UHFFFAOYSA-O
Synonym Methylamine Tetrafluoroborate
PubChem CID 87173277
IUPAC Name methylazanium;tetrafluoroborate
SMILES [B-](F)(F)(F)F.C[NH3+]

Specifications

Melting Point 199°C
Color White
Quantity 1 g
UN Number 1759
Formula Weight 118.87
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Methylammonium Tetrafluoroborate