missing translation for 'onlineSavingsMsg'
Learn More

Methylaminoacetonitrile Sulfate 95.0+%, TCI America™

Supplier:  TCI America M014125G

Catalog No. M014125G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Methylaminoacetonitrile Sulfate
161°C
C3H8N2O4S
25 g
C[NH2+]CC#N.OS([O-])(=O)=O
168.17
238.26
Crystalline Powder
33986-15-1
White-Yellow
MFCD00035599
SEBZVAFMXLJJNO-UHFFFAOYSA-N
(cyanomethyl)(methyl)azanium hydrogen sulfate
44721378
≥95.0% (T)

Chemical Identifiers

33986-15-1
168.17
SEBZVAFMXLJJNO-UHFFFAOYSA-N
(cyanomethyl)(methyl)azanium hydrogen sulfate
C3H8N2O4S
MFCD00035599
44721378
C[NH2+]CC#N.OS([O-])(=O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : AM0875000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions