missing translation for 'onlineSavingsMsg'
Learn More

Methylaminoacetaldehyde Dimethyl Acetal 98.0+%, TCI America™

Catalog No. M0884500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0884500ML 500 mL
M088425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0884500ML Supplier TCI America Supplier No. M0884500ML

Chemical Identifiers

CAS 122-07-6
Molecular Formula C5H13NO2
Molecular Weight (g/mol) 119.164
MDL Number MFCD00008485
InChI Key HUMIEJNVCICTPJ-UHFFFAOYSA-N
Synonym methylaminoacetaldehyde dimethyl acetal, methylamino acetaldehyde dimethyl acetal, ethanamine, 2,2-dimethoxy-n-methyl, 2,2-dimethoxyethyl methyl amine, n-methylaminoacetaldehyde dimethyl acetal, n-2,2-dimethoxyethyl methylamine, unii-6v8q54khcl, 2,2-dimethoxyethylmethylamine, 2-methylamino acetaldehyde dimethyl acetal
PubChem CID 8503
IUPAC Name 2,2-dimethoxy-N-methylethanamine
SMILES CNCC(OC)OC

Specifications

Color Yellow
Boiling Point 140°C
Quantity 500 mL
UN Number 1993
Formula Weight 119.16
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Methylaminoacetaldehyde Dimethyl Acetal
EINECSNumber (2)-3430
TSCA Yes