missing translation for 'onlineSavingsMsg'
Learn More

(Methylamino)acetaldehyde dimethyl acetal, 97%

Catalog No. AAL0239114
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0239114 25 g
AAL0239122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0239114 Supplier Thermo Scientific Chemicals Supplier No. L0239114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 122-07-6 | C5H13NO2 | 119.164 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 122-07-6
Molecular Formula C5H13NO2
Molecular Weight (g/mol) 119.164
MDL Number MFCD00008485
InChI Key HUMIEJNVCICTPJ-UHFFFAOYSA-N
Synonym methylaminoacetaldehyde dimethyl acetal, methylamino acetaldehyde dimethyl acetal, ethanamine, 2,2-dimethoxy-n-methyl, 2,2-dimethoxyethyl methyl amine, n-methylaminoacetaldehyde dimethyl acetal, n-2,2-dimethoxyethyl methylamine, unii-6v8q54khcl, 2,2-dimethoxyethylmethylamine, 2-methylamino acetaldehyde dimethyl acetal
PubChem CID 8503
IUPAC Name 2,2-dimethoxy-N-methylethanamine
SMILES CNCC(OC)OC

Specifications

Melting Point -73°C
Density 0.928
pH 10.9
Boiling Point 139°C to 140°C
Flash Point 29°C (84°F)
Odor Ammonia-like
Refractive Index 1.4115
Quantity 25 g
UN Number UN1993
Beilstein 605322
Sensitivity Air Sensitive
Formula Weight 119.16
Percent Purity 97%
Chemical Name or Material (Methylamino)acetaldehyde dimethyl acetal
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H318-H226-H335-H315
Causes serious eye damage.
Flammable liquid and vapor.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 204-520-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only