missing translation for 'onlineSavingsMsg'
Learn More

Methyl (Z)-3-(p-Toluenesulfonyloxy)but-2-enoate 98.0+%, TCI America™

Catalog No. M30431G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M30431G 1 g
M30435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M30431G Supplier TCI America Supplier No. M30431G
Only null left

Chemical Identifiers

CAS 1029612-18-7
Molecular Formula C12H14O5S
Molecular Weight (g/mol) 270.299
InChI Key QBENOPMVGLDWSB-NTMALXAHSA-N
Synonym (Z)-3-(p-Toluenesulfonyloxy)but-2-enoic Acid Methyl Ester, Methyl (Z)-3-(Tosyloxy)but-2-enoate
PubChem CID 132274876
IUPAC Name methyl (Z)-3-(4-methylphenyl)sulfonyloxybut-2-enoate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OC(=CC(=O)OC)C

Specifications

Melting Point 65°C
Color White-Yellow
Quantity 1 g
Formula Weight 270.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl (Z)-3-(p-Toluenesulfonyloxy)but-2-enoate