missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Yellow, TCI America™
Supplier: TCI America D023125G
Specifications
| Methyl Yellow | |
| 114°C | |
| MFCD00008308 | |
| 2811 | |
| JCYPECIVGRXBMO-UHFFFAOYSA-N | |
| N,N-dimethyl-4-(2-phenyldiazen-1-yl)aniline | |
| 6053 | |
| 225.30 |
| 60-11-7 | |
| C14H15N3 | |
| 25 g | |
| methyl yellow, dimethyl yellow, 4-dimethylamino azobenzene, 4-dimethylaminoazobenzene, solvent yellow 2, sudan yellow, fat yellow, p-dimethylaminoazobenzene, stear yellow jb, sudan yellow gg | |
| CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1 | |
| 225.30 | |
| CHEBI:17903 | |
| Crystalline Powder |
Chemical Identifiers
| 60-11-7 | |
| 225.30 | |
| JCYPECIVGRXBMO-UHFFFAOYSA-N | |
| 6053 | |
| N,N-dimethyl-4-(2-phenyldiazen-1-yl)aniline |
| C14H15N3 | |
| MFCD00008308 | |
| methyl yellow, dimethyl yellow, 4-dimethylamino azobenzene, 4-dimethylaminoazobenzene, solvent yellow 2, sudan yellow, fat yellow, p-dimethylaminoazobenzene, stear yellow jb, sudan yellow gg | |
| CHEBI:17903 | |
| CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1 |
Safety and Handling
EINECSNumber : (5)-3040
RTECSNumber : BX7350000
TSCA : Yes