missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Yellow, pure, indicator grade

CAS: 60-11-7 | C14H15N3 | 225.30 g/mol

Supplier:  Thermo Scientific Chemicals 151401000

 View more versions of this product

Catalog No. AC151401000


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

Methyl Yellow
60-11-7
(CH3)2NC6H4N=NC6H5
4-Dimethylaminoazobenzene, N, N-Dimethyl-4-phenylazoaniline, Solvent Yellow 2
JCYPECIVGRXBMO-UHFFFAOYSA-N
CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1
225.30
CHEBI:17903
225.29
Glass bottle
Powder
111.0°C to 117.0°C
C14H15N3
MFCD00008308
Solubility in water: 13.6 mg/l. Other solubilities: soluble ca. 140g/L in acetone, soluble in benzene, chloroform, diethyl ether,, ethanol, petroleum ether, vegetable oils, alcohol,, ether, strong mineral acids and soils
Authentic
N,N-dimethyl-4-phenyldiazenylaniline
6053
3% max.
Pure
100 g

Chemical Identifiers

60-11-7
225.30
JCYPECIVGRXBMO-UHFFFAOYSA-N
6053
N,N-dimethyl-4-phenyldiazenylaniline
C14H15N3
MFCD00008308
4-Dimethylaminoazobenzene, N, N-Dimethyl-4-phenylazoaniline, Solvent Yellow 2
CHEBI:17903
CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only