missing translation for 'onlineSavingsMsg'
Learn More

Methyl Viologen hydrate, 98%

Catalog No. AC227320050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC227320050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC227320050 Supplier Thermo Scientific Chemicals Supplier No. 227320050
Only null left

CAS: 1910-42-5 | C12H14Cl2N2 | 257.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1910-42-5
Molecular Formula C12H14Cl2N2
Molecular Weight (g/mol) 257.16
MDL Number MFCD00150001
InChI Key FIKAKWIAUPDISJ-UHFFFAOYSA-L
Synonym Paraquat, dichloride
PubChem CID 15938
ChEBI CHEBI:28786
IUPAC Name 1-methyl-4-(1-methylpyridin-1-ium-4-yl)pyridin-1-ium;dichloride
SMILES [Cl-].[Cl-].C[N+]1=CC=C(C=C1)C1=CC=[N+](C)C=C1

Specifications

Melting Point >300.0°C
Assay Percent Range 98%
Solubility Information Solubility in water: very soluble in water.
Infrared Spectrum Authentic
Formula Weight 257.15
Percent Purity 98%
Packaging Glass bottle
Color Beige or Brown to Pink
Quantity 5 g
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Methyl Viologen hydrate, 98%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Fatal in contact with skin.
Very toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 217-615-7
RTECSNumber DW2275000

RUO – Research Use Only