missing translation for 'onlineSavingsMsg'
Learn More

Methyl Violet, Indicator (Basic Violet 1; Methyl Violet 2B), Reagents

Numéro de catalogue. 01336734
Modifier l'affichage
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
01336734 10 g
01336733 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 01336734 Fournisseur Labchem Inc Code fournisseur C611900010C1
Il en reste null

Methyl Violet, Indicator (Basic Violet 1; Methyl Violet 2B), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Spécifications

Chemical Name or Material Methyl Violet, Indicator (Basic Violet 1, Methyl Violet 2B)
CAS 8004-87-3
Molecular Formula C24H27N3.HCl
Synonym Basic Violet 1, N-(4-(bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)methanaminium chloride)
Molecular Weight (g/mol) 393.95
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.0395
Color Blue/Green
Quantity 10 g
Physical Form Solid
Afficher plus de résultats Afficher moins de résultats