missing translation for 'onlineSavingsMsg'
Learn More

Methyl Undecafluorohexanoate 96.0+%, TCI America™

Catalog No. M19135G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M19135G Supplier TCI America Supplier No. M19135G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 424-18-0
Molecular Formula C7H3F11O2
Molecular Weight (g/mol) 328.081
MDL Number MFCD06200802
InChI Key NJXMLQHJFDKLKL-UHFFFAOYSA-N
Synonym Methyl Perfluorohexanoate, Undecafluorohexanoic Acid Methyl Ester, Perfluorohexanoic Acid Methyl Ester
PubChem CID 526464
IUPAC Name methyl 2,2,3,3,4,4,5,5,6,6,6-undecafluorohexanoate
SMILES COC(=O)C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Color Yellow
Boiling Point 122°C
Quantity 5 g
UN Number 1993
Formula Weight 328.08
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Undecafluorohexanoate