missing translation for 'onlineSavingsMsg'
Learn More

Methyl trimethylacetate, 99%

CAS: 598-98-1 | C6H12O2 | 116.16 g/mol

$145.50 - $145.50

Chemical Identifiers

CAS 598-98-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00008843
InChI Key CNMFHDIDIMZHKY-UHFFFAOYSA-N
Synonym methyl pivalate, methyl trimethylacetate, propanoic acid, 2,2-dimethyl-, methyl ester, pivalic acid, methyl ester, unii-bfx9w386ox, bfx9w386ox, pivalic acid methyl ester, methyl pivaloate, methyltrimethylacetate, tert-c4h9cooch3
PubChem CID 69027
IUPAC Name methyl 2,2-dimethylpropanoate
SMILES CC(C)(C)C(=O)OC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC127952500
SDS View Documents
Thermo Scientific Chemicals
127952500
250 mL Glass bottle
Each for $145.50
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

598-98-1
116.16
CNMFHDIDIMZHKY-UHFFFAOYSA-N
69027
CC(C)(C)C(=O)OC
C6H12O2
MFCD00008843
methyl pivalate, methyl trimethylacetate, propanoic acid, 2,2-dimethyl-, methyl ester, pivalic acid, methyl ester, unii-bfx9w386ox, bfx9w386ox, pivalic acid methyl ester, methyl pivaloate, methyltrimethylacetate, tert-c4h9cooch3
methyl 2,2-dimethylpropanoate

Specifications

598-98-1
Colorless
101°C
Authentic
Glass bottle
1.3890 to 1.391
MFCD00008843
02, 320
methyl pivalate, methyl trimethylacetate, propanoic acid, 2,2-dimethyl-, methyl ester, pivalic acid, methyl ester, unii-bfx9w386ox, bfx9w386ox, pivalic acid methyl ester, methyl pivaloate, methyltrimethylacetate, tert-c4h9cooch3
CC(C)(C)C(=O)OC
116.16
30 mPa.s (25°C)
99%
Methyl trimethylacetate
-70°C
0.8700g/mL
<1°C
99%
C6H12O2
(CH3)3CCO2CH3
250 mL
0.87
CNMFHDIDIMZHKY-UHFFFAOYSA-N
methyl 2,2-dimethylpropanoate
69027
116.16
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.

GHS Signal Word: Danger

EINECSNumber : 209-959-1

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only