Learn More
Methyl trimethylacetate, 99%
CAS: 598-98-1 | C6H12O2 | 116.16 g/mol
$217.14 - $217.14
Chemical Identifiers
| CAS | 598-98-1 |
|---|---|
| Molecular Formula | C6H12O2 |
| Molecular Weight (g/mol) | 116.16 |
| MDL Number | MFCD00008843 |
| InChI Key | CNMFHDIDIMZHKY-UHFFFAOYSA-N |
| Synonym | methyl pivalate, methyl trimethylacetate, propanoic acid, 2,2-dimethyl-, methyl ester, pivalic acid, methyl ester, unii-bfx9w386ox, bfx9w386ox, pivalic acid methyl ester, methyl pivaloate, methyltrimethylacetate, tert-c4h9cooch3 |
| PubChem CID | 69027 |
| IUPAC Name | methyl 2,2-dimethylpropanoate |
| SMILES | CC(C)(C)C(=O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1376436
|
Thermo Scientific Chemicals
A1376436 |
500 g |
Each for $217.14
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 598-98-1 | |
| 116.16 | |
| CNMFHDIDIMZHKY-UHFFFAOYSA-N | |
| 69027 | |
| CC(C)(C)C(=O)OC |
| C6H12O2 | |
| MFCD00008843 | |
| methyl pivalate, methyl trimethylacetate, propanoic acid, 2,2-dimethyl-, methyl ester, pivalic acid, methyl ester, unii-bfx9w386ox, bfx9w386ox, pivalic acid methyl ester, methyl pivaloate, methyltrimethylacetate, tert-c4h9cooch3 | |
| methyl 2,2-dimethylpropanoate |
Specifications
| 598-98-1 | |
| 0.875 | |
| 6°C (42°F) | |
| 1.39 | |
| 500 g | |
| 1744141 | |
| CNMFHDIDIMZHKY-UHFFFAOYSA-N | |
| methyl 2,2-dimethylpropanoate | |
| 69027 | |
| 99% |
| <-70°C | |
| 100°C to 101°C | |
| C6H12O2 | |
| MFCD00008843 | |
| UN3272 | |
| methyl pivalate, methyl trimethylacetate, propanoic acid, 2,2-dimethyl-, methyl ester, pivalic acid, methyl ester, unii-bfx9w386ox, bfx9w386ox, pivalic acid methyl ester, methyl pivaloate, methyltrimethylacetate, tert-c4h9cooch3 | |
| CC(C)(C)C(=O)OC | |
| 116.16 | |
| 116.16 | |
| Methyl trimethylacetate |
Safety and Handling
GHS H Statement
H225-H301
Highly flammable liquid and vapour.
Toxic if swallowed.
P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c
H225-H302
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber : 209-959-1
RTECSNumber : UA2459597
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only