missing translation for 'onlineSavingsMsg'
Learn More

Methyl trifluoromethanesulfonate, 97%

CAS: 333-27-7 | C2H3F3O3S | 164.10 g/mol

Supplier:  Thermo Scientific Chemicals H6175822

 View more versions of this product

Catalog No. AAH6175822


Only null left
Explore available promotions

Description

Description

Methyl trifluoromethanesulfonate is used as a powerful methylating reagent in chemistry. Further, it is used in the conversion of amines to methyl ammonium triflates. In addition, it is also used in Suzuki reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl trifluoromethanesulfonate is used as a powerful methylating reagent in chemistry. Further, it is used in the conversion of amines to methyl ammonium triflates. In addition, it is also used in Suzuki reaction.

Notes
Air and moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong bases, strong acids and strong reducing agents. Hydrolyzes with water.
Specifications

Specifications

Methyl trifluoromethanesulfonate
1.5
38°C (100°F)
1.326
MFCD00000409
UN2920
methyl triflate, methyl trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, methyl ester, triflate ester, methyl trifluoromethane sulfonate, trifluoromethanesulfonic acid methyl ester, unii-7b25z22epv, ccris 1158, methyltrifluoromethanesulfonate, methanesulfonic acid, 1,1,1-trifluoro-, methyl ester
COS(=O)(=O)C(F)(F)F
164.10
164.1
333-27-7
99°C
C2H3F3O3S
CF3SO3CH3
100 g
Air sensitive
OIRDBPQYVWXNSJ-UHFFFAOYSA-N
methyl trifluoromethanesulfonate
9526
98%

Chemical Identifiers

333-27-7
164.10
OIRDBPQYVWXNSJ-UHFFFAOYSA-N
9526
COS(=O)(=O)C(F)(F)F
C2H3F3O3S
MFCD00000409
methyl triflate, methyl trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, methyl ester, triflate ester, methyl trifluoromethane sulfonate, trifluoromethanesulfonic acid methyl ester, unii-7b25z22epv, ccris 1158, methyltrifluoromethanesulfonate, methanesulfonic acid, 1,1,1-trifluoro-, methyl ester
methyl trifluoromethanesulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H226-H302+H312+H332-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.

EINECSNumber : 206-371-7

TSCA : Yes

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only