missing translation for 'onlineSavingsMsg'
Learn More

Methyl Trifluoroacetate 98.0+%, TCI America™

Catalog No. T068025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T068025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T068025G Supplier TCI America Supplier No. T068025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 431-47-0
Melting Point -78°C
Color Colorless
Boiling Point 43°C
Molecular Formula C3H3F3O2
Quantity 25 g
MDL Number MFCD00000417
UN Number 2924
Synonym methyl trifluoroacetate, trifluoroacetic acid methyl ester, acetic acid, trifluoro-, methyl ester, methyltrifluoroacetate, 2,2,2-trifluoromethylacetate, acetic acid, 2,2,2-trifluoro-, methyl ester, methyl trifluoracetate, tfamt, methyl triflouroacetate, pubchem12490
InChI Key VMVNZNXAVJHNDJ-UHFFFAOYSA-N
SMILES COC(=O)C(F)(F)F
IUPAC Name methyl 2,2,2-trifluoroacetate
Molecular Weight (g/mol) 128.05
PubChem CID 9893
Formula Weight 128.05
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Trifluoroacetate
Show More Show Less