missing translation for 'onlineSavingsMsg'
Learn More

Methyl trifluoroacetate, 99%, Thermo Scientific™

Catalog No. AC243410250
Change view
Click to view available options
25mL; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC243410250 25mL; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC243410250 Supplier Thermo Scientific Chemicals Supplier No. 243410250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 431-47-0
Density 1.2730g/mL
Boiling Point 43°C to 43.5°C
Flash Point -7°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C3H3F3O2
Refractive Index 1.2890 to 1.291
Linear Formula CF3CO2CH3
Quantity 25mL
MDL Number MFCD00000417
Specific Gravity 1.273
Synonym methyl trifluoroacetate, trifluoroacetic acid methyl ester, acetic acid, trifluoro-, methyl ester, methyltrifluoroacetate, 2,2,2-trifluoromethylacetate, acetic acid, 2,2,2-trifluoro-, methyl ester, methyl trifluoracetate, tfamt, methyl triflouroacetate, pubchem12490
Solubility Information Solubility in water: 8g/L water
InChI Key VMVNZNXAVJHNDJ-UHFFFAOYSA-N
SMILES COC(=O)C(F)(F)F
IUPAC Name methyl 2,2,2-trifluoroacetate
Molecular Weight (g/mol) 128.05
PubChem CID 9893
Viscosity 0.31 mPa.s (20°C)
Formula Weight 128.05
Percent Purity 99%
Chemical Name or Material Methyl trifluoroacetate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-074-5

RUO – Research Use Only