missing translation for 'onlineSavingsMsg'
Learn More

Methyl trifluoroacetate, 98+%

CAS: 431-47-0 | C3H3F3O2 | 128.05 g/mol

Supplier:  Thermo Scientific Chemicals A173380B

 View more versions of this product

Catalog No. AAA173380B


Only null left
Explore available promotions

Description

Description

Useful reagent for trifluoroacetylation of amines under mild conditions. In the presence of 1,1,3,3-Tetramethyl­guanidine, has also been used for the N-acylation of amino acids. It is used as an intermediate in the synthesis of pharmaceutical and agricultural chemicals. The chemical reactions usually involve nucleophilic substitution of the ester and are useful for the preparation of trifluoroacylated compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful reagent for trifluoroacetylation of amines under mild conditions. In the presence of 1,1,3,3-Tetramethyl­guanidine, has also been used for the N-acylation of amino acids. It is used as an intermediate in the synthesis of pharmaceutical and agricultural chemicals. The chemical reactions usually involve nucleophilic substitution of the ester and are useful for the preparation of trifluoroacylated compounds.

Solubility
Not miscible or difficult to mix in water.

Notes
Moisture sensitive. Store away from strong bases, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.
Specifications

Specifications

Methyl trifluoroacetate
-78°C
43°C to 44°C
Pungent
1.29
1000 g
1756070
methyl trifluoroacetate, trifluoroacetic acid methyl ester, acetic acid, trifluoro-, methyl ester, methyltrifluoroacetate, 2,2,2-trifluoromethylacetate, acetic acid, 2,2,2-trifluoro-, methyl ester, methyl trifluoracetate, tfamt, methyl triflouroacetate, pubchem12490
VMVNZNXAVJHNDJ-UHFFFAOYSA-N
methyl 2,2,2-trifluoroacetate
9893
≥98%
431-47-0
1.283
−18°C (−0.4°F)
C3H3F3O2
MFCD00000417
UN2924
Moisture sensitive
Not miscible or difficult to mix in water.
COC(=O)C(F)(F)F
128.05
128.05

Chemical Identifiers

431-47-0
128.05
VMVNZNXAVJHNDJ-UHFFFAOYSA-N
9893
COC(=O)C(F)(F)F
C3H3F3O2
MFCD00000417
methyl trifluoroacetate, trifluoroacetic acid methyl ester, acetic acid, trifluoro-, methyl ester, methyltrifluoroacetate, 2,2,2-trifluoromethylacetate, acetic acid, 2,2,2-trifluoro-, methyl ester, methyl trifluoracetate, tfamt, methyl triflouroacetate, pubchem12490
methyl 2,2,2-trifluoroacetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H225-H314

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

EINECSNumber : 207-074-5

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only