missing translation for 'onlineSavingsMsg'
Learn More

Methyl trifluoroacetate, 98+%

Catalog No. AAA1733830
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1733830 250 g
AAA1733818 50 g
AAA173380B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1733830 Supplier Thermo Scientific Chemicals Supplier No. A1733830
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 431-47-0 | C3H3F3O2 | 128.05 g/mol

Useful reagent for trifluoroacetylation of amines under mild conditions. In the presence of 1,1,3,3-Tetramethyl­guanidine, has also been used for the N-acylation of amino acids. It is used as an intermediate in the synthesis of pharmaceutical and agricultural chemicals. The chemical reactions usually involve nucleophilic substitution of the ester and are useful for the preparation of trifluoroacylated compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful reagent for trifluoroacetylation of amines under mild conditions. In the presence of 1,1,3,3-Tetramethyl­guanidine, has also been used for the N-acylation of amino acids. It is used as an intermediate in the synthesis of pharmaceutical and agricultural chemicals. The chemical reactions usually involve nucleophilic substitution of the ester and are useful for the preparation of trifluoroacylated compounds.

Solubility
Not miscible or difficult to mix in water.

Notes
Moisture sensitive. Store away from strong bases, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.

Chemical Identifiers

CAS 431-47-0
Molecular Formula C3H3F3O2
Molecular Weight (g/mol) 128.05
MDL Number MFCD00000417
InChI Key VMVNZNXAVJHNDJ-UHFFFAOYSA-N
Synonym methyl trifluoroacetate, trifluoroacetic acid methyl ester, acetic acid, trifluoro-, methyl ester, methyltrifluoroacetate, 2,2,2-trifluoromethylacetate, acetic acid, 2,2,2-trifluoro-, methyl ester, methyl trifluoracetate, tfamt, methyl triflouroacetate, pubchem12490
PubChem CID 9893
IUPAC Name methyl 2,2,2-trifluoroacetate
SMILES COC(=O)C(F)(F)F

Specifications

Melting Point -78°C
Density 1.283
Boiling Point 43°C to 44°C
Flash Point −18°C (−0.4°F)
Odor Pungent
Refractive Index 1.29
Quantity 250 g
UN Number UN2924
Beilstein 1756070
Sensitivity Moisture sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 128.05
Percent Purity ≥98%
Chemical Name or Material Methyl trifluoroacetate
Show More Show Less
Hazard Category H225-H314
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 207-074-5
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only