missing translation for 'onlineSavingsMsg'
Learn More

Methyl trans-3-aminocyclohexanecarboxylate hydrochloride, 95%

Catalog No. p-7041410
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62024MD 250 mg
AAH6202403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH62024MD Supplier Thermo Scientific Chemicals Supplier No. H62024MD

CAS: 712313-64-9 | C8H16ClNO2 | 193.67 g/mol

Methyl trans-3-aminocyclohexanecarboxylate hydrochloride used as catalyst in pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl trans-3-aminocyclohexanecarboxylate hydrochloride used as catalyst in pharmaceutical research.

Solubility
Insoluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 712313-64-9
Molecular Formula C8H16ClNO2
Molecular Weight (g/mol) 193.67
MDL Number MFCD12022631,MFCD19237735,MFCD22392158,MFCD31620808,MFCD13184930,MFCD28167966
InChI Key OOFXENVWECZJBF-UHFFFAOYNA-N
Synonym trans-methyl-3-aminocyclohexanecarboxylate hydrochloride, trans-methyl-3-aminocyclohexanecarboxylatehydrochloride, methyl trans-3-aminocyclohexanecarboxylate hydrochloride, methyl 1r,3r-3-aminocyclohexane-1-carboxylate hydrochloride
PubChem CID 46172886
SMILES Cl.COC(=O)C1CCCC(N)C1

Specifications

Solubility Information Insoluble in water.
IUPAC Name methyl (1R,3R)-3-aminocyclohexane-1-carboxylate;hydrochloride
Quantity 250 mg
Formula Weight 193.67
Percent Purity 97%
Chemical Name or Material Methyl trans-3-aminocyclohexanecarboxylate hydrochloride
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only