missing translation for 'onlineSavingsMsg'
Learn More

Methyl trans-2-hexenoate, 97%

Catalog No. p-7044744
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0803009 10 g
AAL0803018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0803009 Supplier Thermo Scientific Chemicals Supplier No. L0803009

CAS: 13894-63-8 | C7H12O2 | 128.171 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13894-63-8
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00048798
InChI Key GFUGBRNILVVWIE-AATRIKPKSA-N
Synonym methyl 2-hexenoate, methyl e-2-hexenoate, methyl trans-2-hexenoate, methyl e-hex-2-enoate, methyl 2e-hexenoate, methyl 2e-hex-2-enoate, 2-hexenoic acid, methyl ester, e, methyl 2e-2-hexenoate, 2e-2-hexenoic acid methyl ester, unii-x2ddu82182
PubChem CID 5364409
IUPAC Name methyl (E)-hex-2-enoate
SMILES CCCC=CC(=O)OC

Specifications

Density 0.915
Boiling Point 57°C to 58°C (13 mmHg)
Flash Point 45°C (113°F)
Odor Fruit-like
Refractive Index 1.4345
Quantity 10 g
UN Number UN3272
Beilstein 1700768
Formula Weight 128.17
Percent Purity 98%
Chemical Name or Material Methyl trans-2-hexenoate
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 237-663-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only