missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Thiophene-3-carboxylate 97.0+%, TCI America™
$91.33 - $363.49
Chemical Identifiers
| CAS | 22913-26-4 |
|---|---|
| Molecular Formula | C6H6O2S |
| Molecular Weight (g/mol) | 142.17 |
| MDL Number | MFCD06203810 |
| InChI Key | ZTRAEMILTFNZSM-UHFFFAOYSA-N |
| Synonym | methyl 3-thiophenecarboxylate, 3-thiophenecarboxylic acid, methyl ester, thiophene-3-carboxylic acid methyl ester, 3-thiophenecarboxylicacid, methyl ester, methyl thiophen-3-carboxylate, methyl-3-thenoate, methyl 3-thenoate, 3-methoxycarbonylthiophene, 3-methoxy carbonyl-thiophene, methyl 3-thiophenecarboxylate # |
| PubChem CID | 574151 |
| IUPAC Name | methyl thiophene-3-carboxylate |
| SMILES | COC(=O)C1=CSC=C1 |
Chemical Identifiers
| 22913-26-4 | |
| 142.17 | |
| ZTRAEMILTFNZSM-UHFFFAOYSA-N | |
| 574151 | |
| COC(=O)C1=CSC=C1 |
| C6H6O2S | |
| MFCD06203810 | |
| methyl 3-thiophenecarboxylate, 3-thiophenecarboxylic acid, methyl ester, thiophene-3-carboxylic acid methyl ester, 3-thiophenecarboxylicacid, methyl ester, methyl thiophen-3-carboxylate, methyl-3-thenoate, methyl 3-thenoate, 3-methoxycarbonylthiophene, 3-methoxy carbonyl-thiophene, methyl 3-thiophenecarboxylate # | |
| methyl thiophene-3-carboxylate |
Specifications
| 22913-26-4 | |
| 91°C | |
| MFCD06203810 | |
| methyl 3-thiophenecarboxylate, 3-thiophenecarboxylic acid, methyl ester, thiophene-3-carboxylic acid methyl ester, 3-thiophenecarboxylicacid, methyl ester, methyl thiophen-3-carboxylate, methyl-3-thenoate, methyl 3-thenoate, 3-methoxycarbonylthiophene, 3-methoxy carbonyl-thiophene, methyl 3-thiophenecarboxylate # | |
| COC(=O)C1=CSC=C1 | |
| 142.17 | |
| 142.17 | |
| Liquid |
| Yellow | |
| C6H6O2S | |
| 5 g | |
| ZTRAEMILTFNZSM-UHFFFAOYSA-N | |
| methyl thiophene-3-carboxylate | |
| 574151 | |
| ≥97.0% (GC) | |
| Methyl Thiophene-3-carboxylate |
Safety and Handling
TSCA : No