missing translation for 'onlineSavingsMsg'
Learn More

Methyl Thiophene-3-carboxylate 97.0+%, TCI America™

Catalog No. M26275G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26275G 5 g
M262725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26275G Supplier TCI America Supplier No. M26275G

Chemical Identifiers

CAS 22913-26-4
Molecular Formula C6H6O2S
Molecular Weight (g/mol) 142.17
MDL Number MFCD06203810
InChI Key ZTRAEMILTFNZSM-UHFFFAOYSA-N
Synonym methyl 3-thiophenecarboxylate, 3-thiophenecarboxylic acid, methyl ester, thiophene-3-carboxylic acid methyl ester, 3-thiophenecarboxylicacid, methyl ester, methyl thiophen-3-carboxylate, methyl-3-thenoate, methyl 3-thenoate, 3-methoxycarbonylthiophene, 3-methoxy carbonyl-thiophene, methyl 3-thiophenecarboxylate #
PubChem CID 574151
IUPAC Name methyl thiophene-3-carboxylate
SMILES COC(=O)C1=CSC=C1

Specifications

Color Yellow
Boiling Point 91°C
Quantity 5 g
Formula Weight 142.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Thiophene-3-carboxylate