missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl thiophene-2-carboxylate, 97%
CAS: 5380-42-7 | C6H6O2S | 142.17 g/mol
Supplier: Thermo Scientific Chemicals A1267722
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Methyl thiophene-2-carboxylate | |
| 1.23 | |
| C6H6O2S | |
| MFCD00016895 | |
| 111111 | |
| PGBFYLVIMDQYMS-UHFFFAOYSA-N | |
| methyl thiophene-2-carboxylate | |
| 79340 | |
| 97% |
| 5380-42-7 | |
| 95°C to 97°C (15 mmHg) | |
| 1.542 | |
| 100 g | |
| methyl 2-thiophenecarboxylate, methyl thenoate, methylthiophene-2-carboxylate, 2-thiophenecarboxylic acid methyl ester, 2-carbomethoxy thiophene, 2-thiophenecarboxylic acid, methyl ester, thiophenate methyl, 2-methoxycarbonyl thiophene, methyl 2-thiophene carboxylate, thiophenate-methyl | |
| COC(=O)C1=CC=CS1 | |
| 142.17 | |
| 142.18 |
Chemical Identifiers
| 5380-42-7 | |
| 142.17 | |
| PGBFYLVIMDQYMS-UHFFFAOYSA-N | |
| 79340 | |
| COC(=O)C1=CC=CS1 |
| C6H6O2S | |
| MFCD00016895 | |
| methyl 2-thiophenecarboxylate, methyl thenoate, methylthiophene-2-carboxylate, 2-thiophenecarboxylic acid methyl ester, 2-carbomethoxy thiophene, 2-thiophenecarboxylic acid, methyl ester, thiophenate methyl, 2-methoxycarbonyl thiophene, methyl 2-thiophene carboxylate, thiophenate-methyl | |
| methyl thiophene-2-carboxylate |
Safety and Handling
EINECSNumber : 226-371-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only