missing translation for 'onlineSavingsMsg'
Learn More

Methyl thiophene-2-carboxylate, 97%

Catalog No. AAA1267706
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1267706 5 g
AAA1267714 25 g
AAA1267722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1267706 Supplier Thermo Scientific Chemicals Supplier No. A1267706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5380-42-7 | C6H6O2S | 142.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5380-42-7
Molecular Formula C6H6O2S
Molecular Weight (g/mol) 142.17
MDL Number MFCD00016895
InChI Key PGBFYLVIMDQYMS-UHFFFAOYSA-N
Synonym methyl 2-thiophenecarboxylate, methyl thenoate, methylthiophene-2-carboxylate, 2-thiophenecarboxylic acid methyl ester, 2-carbomethoxy thiophene, 2-thiophenecarboxylic acid, methyl ester, thiophenate methyl, 2-methoxycarbonyl thiophene, methyl 2-thiophene carboxylate, thiophenate-methyl
PubChem CID 79340
IUPAC Name methyl thiophene-2-carboxylate
SMILES COC(=O)C1=CC=CS1

Specifications

Density 1.23
Boiling Point 95°C to 97°C (15 mmHg)
Refractive Index 1.542
Quantity 5 g
Beilstein 111111
Formula Weight 142.18
Percent Purity 97%
Chemical Name or Material Methyl thiophene-2-carboxylate
EINECSNumber 226-371-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only