missing translation for 'onlineSavingsMsg'
Learn More

Methyl thiazolidine-2-carboxylate hydrochloride, 98%

Catalog No. AAL1074003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1074003 1 g
AAL1074006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1074003 Supplier Thermo Scientific Chemicals Supplier No. L1074003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 50703-06-5 | C5H10ClNO2S | 183.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50703-06-5
Molecular Formula C5H10ClNO2S
Molecular Weight (g/mol) 183.65
MDL Number MFCD00068022
InChI Key YEFOSJYXVLNYSL-UHFFFAOYNA-N
Synonym 2-thiazolidinecarboxylic acid methyl ester, 2-thiazolidinecarboxylic acid, methyl ester, thiazolidine-2-carboxylic acid methyl ester, methyl thiazolidine-2-carboxylate, +-thiazolidine-2-carboxylic acid methyl ester, +/--thiazolidine-2-carboxylic acid methyl ester, 2-thiazolidinecarboxylic acid, methyl ester 6ci,8ci,9ci
PubChem CID 2737507
IUPAC Name methyl 1,3-thiazolidine-2-carboxylate
SMILES [H+].[Cl-].COC(=O)C1CSCN1

Specifications

Melting Point ∽158°C (decomposition)
Quantity 1 g
Beilstein 3628640
Formula Weight 183.66
Percent Purity 98%
Chemical Name or Material Methyl thiazolidine-2-carboxylate hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 256-726-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only