missing translation for 'onlineSavingsMsg'
Learn More

Methyl tetrahydro-4-oxo-2H-thiopyran-3-carboxylate, 95%

Catalog No. AC434290050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434290050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC434290050 Supplier Thermo Scientific Chemicals Supplier No. 434290050

CAS: 4160-61-6 | C7H10O3S | 174.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4160-61-6
Molecular Formula C7H10O3S
Molecular Weight (g/mol) 174.22
InChI Key MCUXKFHPGMEIIW-UHFFFAOYSA-N
Synonym methyl 4-oxotetrahydro-2h-thiopyran-3-carboxylate, methyl tetrahydro-4-oxo-2h-thiopyran-3-carboxylate, methyl 4-oxotetrahydrothiopyran-3-carboxylate, 2h-thiopyran-3-carboxylic acid, tetrahydro-4-oxo-, methyl ester, methyl4-oxotetrahydro-2h-thiopyran-3-carboxylate, methyl-4-oxotetrahydro-2h-thiopyran-3-carboxylate, 2,3,5,6-tetrahydro-3-carbomethoxythiopyran-4-one, 4-oxotetrahydro-2h-thiopyran-3-carbonsauremethylester, 4-oxotetrahydro-2h-thiopyran-3-carboxylic acid methyl ester, 4-oxo-tetrahydro-thiopyran-3-carboxylic acid methyl ester
PubChem CID 10374916
IUPAC Name methyl 4-oxothiane-3-carboxylate
SMILES COC(=O)C1CSCCC1=O

Specifications

Color Colorless to Yellow
Boiling Point 134°C (10.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 174.22
Percent Purity 95%
Physical Form Liquid or Solid
Chemical Name or Material Methyl tetrahydro-4-oxo-2H-thiopyran-3-carboxylate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only