missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Terephthalaldehydate 98.0+%, TCI America™
Supplier: TCI America T0012100G
Specifications
| Methyl Terephthalaldehydate | |
| 60°C | |
| 135°C | |
| MFCD00006950 | |
| methyl p-formylbenzoate, 4-carbomethoxybenzaldehyde, methyl terephthalaldehydate, 4-methoxycarbonyl benzaldehyde, benzoic acid, 4-formyl-, methyl ester, p-carbomethoxybenzaldehyde, methyl terephthaldehydate, methyl4-formylbenzoate, 4-formylbenzoic acid methyl ester, methyl-p-formyl benzoate | |
| COC(=O)C1=CC=C(C=O)C=C1 | |
| 164.16 | |
| 164.16 | |
| Crystalline Powder |
| 1571-08-0 | |
| White | |
| C9H8O3 | |
| 100 g | |
| FEIOASZZURHTHB-UHFFFAOYSA-N | |
| methyl 4-formylbenzoate | |
| 15294 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 1571-08-0 | |
| 164.16 | |
| FEIOASZZURHTHB-UHFFFAOYSA-N | |
| 15294 | |
| COC(=O)C1=CC=C(C=O)C=C1 |
| C9H8O3 | |
| MFCD00006950 | |
| methyl p-formylbenzoate, 4-carbomethoxybenzaldehyde, methyl terephthalaldehydate, 4-methoxycarbonyl benzaldehyde, benzoic acid, 4-formyl-, methyl ester, p-carbomethoxybenzaldehyde, methyl terephthaldehydate, methyl4-formylbenzoate, 4-formylbenzoic acid methyl ester, methyl-p-formyl benzoate | |
| methyl 4-formylbenzoate |
Safety and Handling
EINECSNumber : (3)-1661
TSCA : Yes