missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl t-Butyl Ether, for HPLC, GC, pesticide residue analysis and spectrophotometry, >99.0%, Solstice
Specifications
Specifications
| Absorbance | 0.5 AU max. at 225 nm, 0.1 AU max. at 250 nm |
| CAS Max % | 100.0000% |
| Density | 0.74 g/cm3 |
| Quantity | 6 x 1 L |
| Flash Point | −27°C |
| Packaging | Glass Bottle |
| UN Number | UN2398 |
| Beilstein | 1730942 |
| Boiling Point | 55.2°C |
| Formula Weight | 88.14 |
| Show More |
Safety and Handling
| ShelfLife | 547 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Methyl tert-butyl ether |