missing translation for 'onlineSavingsMsg'
Learn More

Methyl-t-butyl ether, 99.0% min., ACS, MilliporeSigma™

Catalog No. MMX08193
Change view
Click to view available options
Quantity:
4 L
200 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MMX08193 4 L
MMX081930 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MMX08193 Supplier Technidata Supplier No. MX08193

Chemical Identifiers

CAS 1634-04-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
Synonym tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
PubChem CID 15413
ChEBI CHEBI:27642
IUPAC Name 2-methoxy-2-methylpropane
SMILES CC(C)(C)OC

Specifications

Quantity 4 L
Physical Form Liquid
Assay Percent Range 99.0%, min.
Linear Formula CH3OC(CH3)3
Formula Weight 88.14
Grade ACS
Chemical Name or Material Methyl-t-butyl ether, For ACS analysis
Recommended Storage Room Temperature