missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl sulfoxide, 99.7+%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 348440010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Methyl sulfoxide | |
| Brown to White | |
| 1.1000g/mL | |
| 1 L | |
| Authentic | |
| Powder, Needles or Crystals | |
| 99.7+% | |
| C2H6OS | |
| MFCD00002089 | |
| 0.005% Max. | |
| 01,296; 02,157; 03,119; 04,192; 05,263; 06,225; 07,133; 08,198; 09,189; 10,166; 12,212; 13,124; 15,146; 16,149; 17,127 | |
| dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
| CS(C)=O | |
| 78.13 | |
| CHEBI:28262 | |
| ≥99.7% |
| 18.4°C | |
| >190°C | |
| 189.0°C | |
| 87°C | |
| AcroSeal™ Glass Bottle | |
| 1.4770 to 1.4800 | |
| 1.1 | |
| (CH3)2SO | |
| 0.55mbar at 20°C | |
| 01, IV, 1277 | |
| 15, 3285 | |
| IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
| methylsulfinylmethane | |
| 679 | |
| 78.13 | |
| Extra Dry over Molecular Sieve |
Chemical Identifiers
| C2H6OS | |
| MFCD00002089 | |
| dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
| CHEBI:28262 | |
| CS(C)=O |
| 78.13 | |
| IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
| 679 | |
| methylsulfinylmethane |
RUO – Research Use Only