missing translation for 'onlineSavingsMsg'
Learn More

Methyl (S)-(-)-2-Isocyanato-4-methylvalerate 98.0+%, TCI America™

Catalog No. I04675G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04675G 5 g
I046725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I04675G Supplier TCI America Supplier No. I04675G
Only null left

Chemical Identifiers

CAS 39570-63-3
Molecular Formula C8H13NO3
Molecular Weight (g/mol) 171.196
MDL Number MFCD00191532
InChI Key UWFLIWPVRTVDNO-ZETCQYMHSA-N
Synonym (S)-(-)-2-Isocyanato-4-methylvaleric Acid Methyl Ester
PubChem CID 7019163
IUPAC Name methyl (2S)-2-isocyanato-4-methylpentanoate
SMILES CC(C)CC(C(=O)OC)N=C=O

Specifications

Color Yellow
Boiling Point 74°C
Quantity 5 g
UN Number 2206
Formula Weight 171.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl (S)-(-)-2-Isocyanato-4-methylvalerate