missing translation for 'onlineSavingsMsg'
Learn More

Methyl Red 92.0+%, TCI America™

Catalog No. M070225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M070225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M070225G Supplier TCI America Supplier No. M070225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 493-52-7
Molecular Formula C15H15N3O2
Molecular Weight (g/mol) 269.304
MDL Number MFCD00002425
InChI Key CEQFOVLGLXCDCX-UHFFFAOYSA-N
Synonym methyl red, o-methyl red, c.i. acid red 2, acid red 2, cerven methylova, cerven kysela 2, 4-dimethylamino-2'-carboxylazobenzene, 2-carboxy-4'-dimethylamino azobenzene, 2-4-dimethylaminophenylazo benzoic acid, ci acid red 2
PubChem CID 10303
IUPAC Name 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O

Specifications

Melting Point 182°C
Quantity 25 g
Formula Weight 269.30
Percent Purity ≥92.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Red
RTECSNumber DG8960000
TSCA Yes