missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Red, pure, indicator

Catalog No. p-4259091
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC264440250 25 g Glass bottle
AC264441000 100 g Glass bottle
AC264445000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC264440250 Supplier Thermo Scientific Chemicals Supplier No. 264440250
Only null left

Methyl Red, C15H15N3O2, CAS Number-493-52-7, Acid Red 2, 2-[4-(Dimethylamino)phenylazo]benzoic acid, C.I. 13020, 25g, 269.3, Pure, Warning, GHS H Statement: Harmful if swallowed. | CAS: 493-52-7 | C15H15N3O2 | 269.304 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 493-52-7
Molecular Formula C15H15N3O2
Molecular Weight (g/mol) 269.304
InChI Key CEQFOVLGLXCDCX-UHFFFAOYSA-N
Synonym Acid Red 2, 2-[4-(Dimethylamino)phenylazo]benzoic acid, C.I. 13020
PubChem CID 10303
IUPAC Name 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O

Specifications

Melting Point 178.0°C to 182.0°C
Linear Formula (CH3)2NC6H4N=NC6H4CO2H
Solubility Information Solubility in water: almost insoluble. Other solubilities: soluble in ethanol, acetic acid
Infrared Spectrum Authentic
Formula Weight 269.3
Grade Pure
Packaging Glass bottle
Specific Extinction 650 min. (430nm, in buffer)
Visual Transition Interval from pH 4.2 (red) to pH 6.2 (yellow)
Color Red
Quantity 25 g
Physical Form Powder
Chemical Name or Material Methyl Red
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only