missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Red Indicator, 0.02% (w/v) in Methanol, Ricca Chemical
$151.00 - $198.00
Specifications
| Chemical Name or Material | Methyl Red Indicator |
|---|---|
| CAS | 67-56-1 , 493-52-7 |
| Percent Purity | 0.02% W/V |
| Grade | Indicator |
| Concentration or Composition (by Analyte or Components) | 0.02% (w/v) in Methanol |