missing translation for 'onlineSavingsMsg'
Learn More

Methyl Red Indicator, 0.02% Aqueous, Reagents

Catalog No. 01336549
Change view
Click to view available options
Quantity:
125 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336549 125 mL
01336548 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01336549 Supplier Labchem Inc Supplier No. C7118600125C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Methyl Red Indicator, 0.02% Aqueous, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Methyl Red Indicator, 0.02% Aqueous
CAS 7732-18-5, 845-10-3
Boiling Point 100°C
Packaging Glass Bottle
Specific Gravity 1.001
Synonym Acid Red 2, 2-[[4-(Dimethylamino)phenyl]diazenyl]benzoic Acid
Color Orange to Red
Quantity 125 mL
Physical Form Liquid
Percent Purity 0.02%
Grade Reagent
Show More Show Less