missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Red hydrochloride, ACS reagent

CAS: 63451-28-5 | C15H16ClN3O2 | 305.762 g/mol

$65.79

Chemical Identifiers

CAS 63451-28-5
Molecular Formula C15H16ClN3O2
Molecular Weight (g/mol) 305.762
MDL Number MFCD00012614
InChI Key ICTWKXAVJAHRMK-UHFFFAOYSA-N
Synonym Acid Red 2, 2-[4-(Dimethylamino)phenylazo]benzoic acid hydrochloride, C.I. 13020
PubChem CID 113227
IUPAC Name 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid;hydrochloride
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O.Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC339320250
View Documents Promotion Details
Thermo Scientific Chemicals
339320250
25 g Glass bottle
Each for $65.79
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

63451-28-5
305.762
ICTWKXAVJAHRMK-UHFFFAOYSA-N
113227
CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O.Cl
C15H16ClN3O2
MFCD00012614
Acid Red 2, 2-[4-(Dimethylamino)phenylazo]benzoic acid hydrochloride, C.I. 13020
2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid;hydrochloride

Specifications

63451-28-5
C15H16ClN3O2
MFCD00012614
15, 6192
ICTWKXAVJAHRMK-UHFFFAOYSA-N
Authentic
305.762
305.76
from pH 4.2 (pink) to pH 6.2 (yellow)
Powder
Passes Test
(CH3)2NC6H4NNC6H4COOH·HCl
16, 329
Acid Red 2, 2-[4-(Dimethylamino)phenylazo]benzoic acid hydrochloride, C.I. 13020
CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O.Cl
2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid;hydrochloride
113227
ACS Reagent
Black to Blue
Methyl Red hydrochloride
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 264-190-9

SDS
Documents

Documents

RUO – Research Use Only