missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Methyl Red hydrochloride, ACS
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 175°C (decomposition) |
| Quantity | 100 g |
| Physical Form | Crystalline |
| Formula Weight | 305.77 |
| Grade | ACS |
| Chemical Name or Material | Methyl Red hydrochloride |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H351 Suspected of causing cancer. |
| Precautionary Statement | GHS P Statement P281-P201-P202-P308+P313-P405-P501a Use personal protective equipment as required. Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. IF exposed or concerned: Get medical advice/attention. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| EINECSNumber | 264-190-9 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only