missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Red hydrochloride, ACS

Catalog No. AA3666814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3666814 25 g
AA3666822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3666814 Supplier Thermo Scientific Chemicals Supplier No. 03666814
Only null left

CAS: 63451-28-5 | C15H16ClN3O2 | 305.762 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 63451-28-5
Molecular Formula C15H16ClN3O2
Molecular Weight (g/mol) 305.762
MDL Number MFCD00012614
InChI Key ICTWKXAVJAHRMK-UHFFFAOYSA-N
Synonym 2-[4-(Dimethylamino)phenyl]azobenzoic acid
PubChem CID 113227
IUPAC Name 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid;hydrochloride
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O.Cl

Specifications

Melting Point 175°C (decomposition)
Formula Weight 305.77
Grade ACS
Quantity 25 g
Physical Form Crystalline
Chemical Name or Material Methyl Red hydrochloride
Hazard Category Warning
Hazard Statement GHS H Statement
H351
Suspected of causing cancer.
Precautionary Statement GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 264-190-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only